Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF-1/NR5A1/Steroidogenic Factor 1 Rabbit anti-Human, Mouse, Rat, Clone: 3G7Z3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | SF-1/NR5A1/Steroidogenic Factor 1 |
---|---|
Clone | 3G7Z3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
SF-1/NR5A1/Steroidogenic Factor 1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2516 | |
IgG | |
Affinity purified |
3G7Z3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 351-461 of human SF-1/NR5A1/Steroidogenic Factor 1 (Q13285). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title