Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SF Broth

Catalog No. r064532
Click to view available options
Quantity:
20 x 5 mL
This item is not returnable. View return policy

Catalog No. R064532

Supplier: Thermo Scientific™ R064532

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Differentiate enterococci from group D streptococci and other Streptococcus species with Thermo Scientific™ Remel™ SF Broth.

Differentiate enterococci from group D streptococci and other Streptococcus species with Thermo Scientific™ Remel™ SF Broth. SF Broth was developed by Hajna and Perry to detect fecal streptococci in water, milk and other materials of sanitary importance1. Snyder and Lichstein had earlier demonstrated that sodium azide prevented swarming of Proteus and allows selective isolation of fecal streptococci from clinical samples. Fecal streptococci consist of enterococci and group D streptococci of human origin that usually occur in higher densities in animal feces2.

SF Broth is a liquid medium that is used to differentiate enterococci from group D streptococci and other Streptococcus species.

Enterococci grow in SF Broth and produce an acid (yellow) reaction, while other streptococci fail to grow and the medium remains purple in color.

  • Ready-to-use – Convenience of prepared media.
  • Bacteriostatic effect – Sodium azide is an inhibitor that exhibits bacteriostatic effect on gram-negative bacteria.
  • Easy-to-interpret – Turbidity and a yellow, yellow-brown color due to fermentation of dextrose indicates a positive test. Negative test shows lack of turbidity and no color change i.e the medium remains purple.

SF Broth contains tryptone, dextrose, sodium chloride, dipotassium phosphate, monopotassium phosphate, sodium azide, brom cresol purple and demineralized water. Tryptone and dextrose supply nutrients necessary to support the growth of streptococci. Dextrose is the fermentable carbohydrate. Sodium azide inhibits the cytochrome oxidase enzyme in the electron transport chain, thus exhibiting a bacteriostatic effect on gram-negative bacteria. Brom cresol purple is the pH indicator.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description SF Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.