Learn More
SF1 Mouse anti-Human, Clone: OTI7H9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809477

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Steroidogenic factor-1 (SF-1) regulates multiple genes involved in the adrenal and gonadal development and in the biosynthesis of a variety of hormones, including adrenal and gonadal steroids, anti-Mullerian hormone (AMH), and gonadotropins. SF-1 belongs to the fushi tarazu factor-1 (FTZ-F1) subfamily of orphan nuclear receptors. In the adult ovary, SF-1 localizes to theca/interstitial cells.Specifications
SF1 | |
Monoclonal | |
Unconjugated | |
SF1 | |
BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7536 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7H9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15637 | |
SF1 | |
Human recombit protein fragment corresponding to amino acids 1-276 of human SF1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.