Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF20/MYDGF Antibody (2562A) [Allophycocyanin], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB1104A
Description
SF20/MYDGF Monoclonal antibody specifically detects SF20/MYDGF in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
SF20/MYDGF | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
2562A | |
Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10 | |
Mouse myeloma cell line NS0-derived mouse SF20/MYDGF, Val25-Leu166, Accession # Q9CPT4 | |
0.1 mL | |
Biologically Active Proteins, Innate Immunity | |
56005 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction