Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF3A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP187214
Description
SF3A1 Polyclonal specifically detects SF3A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
SF3A1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
pre-mRNA processing 21, pre-mRNA splicing factor SF3a (120 kDa subunit), Prp21, PRPF21, SAP 114, SAP114splicing factor 3a, subunit 1, 120kD, SF3a120, spliceosome associated protein 114, Spliceosome-associated protein 114, splicing factor 3 subunit 1, splicing factor 3A subunit 1, splicing factor 3a, subunit 1, 120kDa | |
Rabbit | |
Affinity Purified | |
RUO | |
10291 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
SF3A1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TEDSLMPEEEFLRRNKGPVSIKVQVPNMQDKTEWKLNGQVLVFTLPLTDQVSVIKVKIHEATGMPAGKQKLQYEGIFIKDSNSLAYYNMANGAVIHLALKESGGRKK | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction