Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF3A1 Antibody (OTI3F4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274150F
Description
SF3A1 Monoclonal specifically detects SF3A1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
SF3A1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SF3A1 | |
Recombinant protein fragment corresponding to amino acids 249-568 of human SF3A1 (NP_005868) produced in E.coli. | |
0.1 mL | |
10291.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI3F4 | |
Immunohistochemistry | |
pre-mRNA processing 21, pre-mRNA splicing factor SF3a (120 kDa subunit), Prp21, PRPF21, SAP 114, SAP114splicing factor 3a, subunit 1, 120kD, SF3a120, spliceosome associated protein 114, Spliceosome-associated protein 114, splicing factor 3 subunit 1, splicing factor 3A subunit 1, splicing factor 3a, subunit 1, 120kDa | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction