Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF3A1 Monoclonal Antibody (OTI4D8)
GREENER_CHOICE

Catalog No. PIMA526468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526468 Supplier Invitrogen™ Supplier No. MA526468
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SF3A1 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes subunit 1 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 1 belongs to the SURP protein family; named for the SURP motifs that are thought to mediate RNA binding. Subunit 1 has tandemly repeated SURP motifs in its amino-terminal half while its carboxy-terminal half contains a proline-rich region and a ubiquitin-like domain. Binding studies with truncated subunit 1 derivatives demonstrated that the two SURP motifs are necessary for binding to subunit 3 while contacts with subunit 2 may occur through sequences carboxy-terminal to the SURP motifs. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF3A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Sf3a1
Gene Accession No. Q15459
Gene Alias 1200014H24Rik; 5930416L09Rik; AI159724; pre-mRNA processing 21; pre-mRNA splicing factor SF3a (120 kDa subunit); PRP21; PRPF21; SAP 114; SAP114; Sf3a1; sf3a1 protein; SF3A120; si:dz150f13.2; spliceosome-associated protein 114; splicing factor 3 subunit 1; splicing factor 3A subunit 1; splicing factor 3a, subunit 1; splicing factor 3a, subunit 1, 120kD; splicing factor 3a, subunit 1, 120kDa; Unknown (protein for MGC:65786); wu:fc38e01; wu:fc50a11; wu:fj37c05; zgc:65786
Gene Symbols Sf3a1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 249-568 of SF3A1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10291
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.