Learn More
SF3A1 Mouse anti-Human, Clone: OTI2C4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800587

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes subunit 1 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 1 belongs to the SURP protein family; named for the SURP motifs that are thought to mediate RNA binding. Subunit 1 has tandemly repeated SURP motifs in its amino-terminal half while its carboxy-terminal half contains a proline-rich region and a ubiquitin-like domain. Binding studies with truncated subunit 1 derivatives demonstrated that the two SURP motifs are necessary for binding to subunit 3 while contacts with subunit 2 may occur through sequences carboxy-terminal to the SURP motifs. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
SF3A1 | |
Monoclonal | |
Unconjugated | |
SF3A1 | |
PRP21, PRPF21, SAP114, SF3A120 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10291 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15459 | |
SF3A1 | |
Human recombit protein fragment corresponding to amino acids 249-568 of human SF3A1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.