Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF3A1 Polyclonal Antibody, Huabio

Catalog No. 502544590
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4590 Supplier HUABIO RESEARCH INC Supplier No. HA500175
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SF3A1 Polyclonal antibody specifically detects SF3A1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SF3A1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Sf3a1
Gene Accession No. Q15459, Q8K4Z5
Gene Alias 1200014H24Rik; 5930416L09Rik; AI159724; pre-mRNA processing 21; pre-mRNA splicing factor SF3a (120 kDa subunit); PRP21; PRPF21; SAP 114; SAP114; Sf3a1; sf3a1 protein; SF3A120; si:dz150f13.2; spliceosome-associated protein 114; splicing factor 3 subunit 1; splicing factor 3A subunit 1; splicing factor 3a, subunit 1; splicing factor 3a, subunit 1, 120kD; splicing factor 3a, subunit 1, 120kDa; Unknown (protein for MGC:65786); wu:fc38e01; wu:fc50a11; wu:fj37c05; zgc:65786
Host Species Rabbit
Immunogen Synthetic peptide within human SF3A1 aa 744-793.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10291, 67465
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.