Learn More
Invitrogen™ SF3A2 Polyclonal Antibody

Description
Immunogen sequence: EVKKFVKIGR PGYKVTKQRD SEMGQQSLLF QIDYPEIAEG IMPRHRFMSA YEQRIEPPDR RWQYLLMAAE PYETIAFKVP SREIDKAEGK FWTHWNRETK QFFLQFHFKM EKP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
Specifications
Specifications
| Antigen | SF3A2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SF3A2 |
| Gene Accession No. | Q15428 |
| Gene Alias | 66kDa; AW047242; pre-mRNA splicing factor SF3A, subunit 2; PRP11; PRPF11; SAP 62; Sap62; Sf3a2; SF3a66; SFA66; spliceosomal protein gene 62; spliceosome associated protein 62; spliceosome-associated protein 62; Splicing factor 3A subunit 2; splicing factor 3a, subunit 2; splicing factor 3a, subunit 2, 66kD; splicing factor 3a, subunit 2, 66kDa |
| Gene Symbols | SF3A2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.