Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SF3A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561969
Description
Immunogen sequence: SSSESNRDRR ERLRQLALET IDINKDPYFM KNHLGSYECK LCLTLHNNEG SYLAHTQGKK HQTNLARRAA KEAKEAPAQP APEKVKVE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes subunit 2 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 2 interacts with subunit 1 through its amino-terminus while the single zinc finger domain of subunit 2 plays a role in its binding to the 15S U2 snRNP. Subunit 2 may also function independently of its RNA splicing function as a microtubule-binding protein.
Specifications
SF3A2 | |
Polyclonal | |
Unconjugated | |
SF3A2 | |
66kDa; AW047242; pre-mRNA splicing factor SF3A, subunit 2; PRP11; PRPF11; SAP 62; Sap62; Sf3a2; SF3a66; SFA66; spliceosomal protein gene 62; spliceosome associated protein 62; spliceosome-associated protein 62; Splicing factor 3A subunit 2; splicing factor 3a, subunit 2; splicing factor 3a, subunit 2, 66kD; splicing factor 3a, subunit 2, 66kDa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15428 | |
SF3A2 | |
Recombinant protein corresponding to Human SF3A2. Recombinant protein control fragment (Product #RP-98921). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction