Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF3A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588047 Supplier Invitrogen™ Supplier No. PA588047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDFQHRPGGK TGSGGVASSS ESNRDRRERL RQLALETIDI NKDPYFMKNH LGSYECKLCL TLHNNEGSYL AHTQGKKHQT NLARRAAKEA KEAPAQPAPE KVKVEVKKFV KIGRPGYKVT KQRDSEMGQQ SLLFQIDYPE IAEGIMPRHR FMSAYEQRIE PPDRRWQYLL MAAEPYETIA FKVPSREIDK AEGKFWTHWN RETKQFFLQF HFKMEKPPAP; Positive Samples: Mouse testis, Rat testis; Cellular Location: Nucleus.

This gene encodes subunit 2 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 2 interacts with subunit 1 through its amino-terminus while the single zinc finger domain of subunit 2 plays a role in its binding to the 15S U2 snRNP. Subunit 2 may also function independently of its RNA splicing function as a microtubule-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF3A2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SF3A2
Gene Accession No. Q62203, Q6AXT8
Gene Alias 66kDa; AW047242; pre-mRNA splicing factor SF3A, subunit 2; PRP11; PRPF11; SAP 62; Sap62; Sf3a2; SF3a66; SFA66; spliceosomal protein gene 62; spliceosome associated protein 62; spliceosome-associated protein 62; Splicing factor 3A subunit 2; splicing factor 3a, subunit 2; splicing factor 3a, subunit 2, 66kD; splicing factor 3a, subunit 2, 66kDa
Gene Symbols SF3A2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human SF3A2 (NP_0090962).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20222, 299620
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.