Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF3A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | SF3A3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SF3A3 Polyclonal specifically detects SF3A3 in Human samples. It is validated for Western Blot.Specifications
| SF3A3 | |
| Polyclonal | |
| Rabbit | |
| PRP9, PRPF9, SAP 61, SAP61splicing factor 3a, subunit 3, 60kD, SF3a60pre-mRNA splicing factor SF3a (60kD), spliceosome associated protein 61, Spliceosome-associated protein 61, splicing factor 3A subunit 3, splicing factor 3a, subunit 3, 60kDa | |
| SF3A3 | |
| IgG | |
| This product is specific to Subunit or Isoform: 3. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 10946 | |
| Synthetic peptides corresponding to SF3A3(splicing factor 3a, subunit 3, 60kDa) The peptide sequence was selected from the middle region of SF3A3. Peptide sequence THENVQRKQARTGEEREEEEEEQISESESEDEENEIIYNPKNLPLGWDGK. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title