Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF3A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556909 Supplier Invitrogen™ Supplier No. PA556909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGWDGKPIPY WLYKLHGLNI NYNCEICGNY TYRGPKAFQR HFAEWRHAHG MRCLGIPNTA HFANVTQIED AVSLWAKLKL QK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP. This gene has a pseudogene on chromosome 20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF3A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SF3A3
Gene Accession No. Q12874
Gene Alias 4930512K19Rik; 60kDa; hypothetical protein LOC523250; LRRGT00093; pre-mRNA splicing factor SF3a (60kD); PRP9; PRPF9; SAP 61; Sap61; SF3A3; SF3a60; spliceosome associated protein 61; spliceosome-associated protein 61; splicesome-associated factor 61; splicing factor 3A subunit 3; splicing factor 3a subunit 3-like protein; splicing factor 3a, subunit 3; splicing factor 3a, subunit 3, 60kD; splicing factor 3a, subunit 3, 60kDa; wu:fi15a06; wu:fj94f01; zgc:110227
Gene Symbols SF3A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SF3A3. Recombinant protein control fragment (Product #RP-102613).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10946
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.