Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF3B14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | SF3B14 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SF3B14 Polyclonal specifically detects SF3B14 in Human samples. It is validated for Western Blot.Specifications
| SF3B14 | |
| Polyclonal | |
| Rabbit | |
| Q9Y3B4 | |
| 51639 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| HSPC175, Ht006, P14, pre-mRNA branch site protein p14, SAP14, SF3b 14 kDa subunit, SF3B14a, spliceosome-associated protein, 14 kDa subunit, splicing factor 3B, 14 kDa subunit | |
| Synthetic peptides corresponding to SF3B14(splicing factor 3B, 14 kDa subunit) The peptide sequence was selected from the N terminal of SF3B14. Peptide sequence MAMQAAKRANIRLPPEVNRILYIRNLPYKITAEEMYDIFGKYGPIRQIRV. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title