Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SF3B14 Monoclonal Antibody (OTI8A1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527406
Description
SF3B14 Monoclonal Antibody for Western Blot, IHC (P)
SF3B14 encodes a 14 kDa protein subunit of the splicing factor 3b complex. Splicing factor 3b associates with both the U2 and U11/U12 small nuclear ribonucleoprotein complexes (U2 snRNP) of spliceosomes. This 14 kDa protein interacts directly with subunit 1 of the splicing factor 3b complex. This 14 kDa protein also interacts directly with the adenosine that carries out the first transesterification step of splicing at the pre-mRNA branch site.
Specifications
SF3B14 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y3B4 | |
SF3B6 | |
Full length human recombinant protein of SF3B14 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI8A1 | |
Unconjugated | |
SF3B6 | |
0610009D07Rik; 6030419K15Rik; AV001342; CGI-110; HSPC175; Ht006; P14; pre mRNA branch site protein p14; pre-mRNA branch site protein p14; SAP14; SAP14a; SF3b 14 kDa subunit; Sf3b14; SF3B14a; SF3B6; spliceosome-associated protein, 14 kDa subunit; spliceosome-associated protein, 14-kDa; Splicing factor 3B subunit 6; splicing factor 3B, 14 kDa subunit; splicing factor 3B, 14 kDa subunit-like; splicing factor 3B, subunit 6; splicing factor 3b, subunit 6, 14kDa | |
Mouse | |
Affinity chromatography | |
RUO | |
51639 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction