Learn More
Invitrogen™ SF3B14 Polyclonal Antibody

Description
Immunogen sequence: YEDIFDAKNA CDHLSGFNVC NRYLVVLYYN ANRAFQKMDT KKKEEQLKLL KEKYGINTDP P Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 70%.
Specifications
Specifications
| Antigen | SF3B14 |
| Applications | Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SF3B6 |
| Gene Accession No. | Q9Y3B4 |
| Gene Alias | 0610009D07Rik; 6030419K15Rik; AV001342; CGI-110; HSPC175; Ht006; P14; pre mRNA branch site protein p14; pre-mRNA branch site protein p14; SAP14; SAP14a; SF3b 14 kDa subunit; Sf3b14; SF3B14a; SF3B6; spliceosome-associated protein, 14 kDa subunit; spliceosome-associated protein, 14-kDa; Splicing factor 3B subunit 6; splicing factor 3B, 14 kDa subunit; splicing factor 3B, 14 kDa subunit-like; splicing factor 3B, subunit 6; splicing factor 3b, subunit 6, 14kDa |
| Gene Symbols | SF3B6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.