Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SF3B14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557078
Description
Immunogen sequence: MAMQAAKRAN IRLPPEVNRI LYIRNLPYKI TAEEMYDIFG KYGPIRQIRV GNTPETRGTA YVV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 95%.
SF3B14 encodes a 14 kDa protein subunit of the splicing factor 3b complex. Splicing factor 3b associates with both the U2 and U11/U12 small nuclear ribonucleoprotein complexes (U2 snRNP) of spliceosomes. This 14 kDa protein interacts directly with subunit 1 of the splicing factor 3b complex. This 14 kDa protein also interacts directly with the adenosine that carries out the first transesterification step of splicing at the pre-mRNA branch site.
Specifications
SF3B14 | |
Polyclonal | |
Unconjugated | |
SF3B6 | |
0610009D07Rik; 6030419K15Rik; AV001342; CGI-110; HSPC175; Ht006; P14; pre mRNA branch site protein p14; pre-mRNA branch site protein p14; SAP14; SAP14a; SF3b 14 kDa subunit; Sf3b14; SF3B14a; SF3B6; spliceosome-associated protein, 14 kDa subunit; spliceosome-associated protein, 14-kDa; Splicing factor 3B subunit 6; splicing factor 3B, 14 kDa subunit; splicing factor 3B, 14 kDa subunit-like; splicing factor 3B, subunit 6; splicing factor 3b, subunit 6, 14kDa | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51639 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3B4 | |
SF3B6 | |
Recombinant protein corresponding to Human SF3B6. Recombinant protein control fragment (Product #RP-96002). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction