Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF3B14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588980 Supplier Invitrogen™ Supplier No. PA588980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAMQAAKRAN IRLPPEVNRI LYIRNLPYKI TAEEMYDIFG KYGPIRQIRV GNTPETRGTA YVVYEDIFDA KNACDHLSGF NVCNRYLVVL YYNANRAFQK MDTKKKEEQL KLLKEKYGIN TDPPK; Positive Samples: K-562, HeLa, Mouse spleen; Cellular Location: Nucleus.

SF3B14 encodes a 14 kDa protein subunit of the splicing factor 3b complex. Splicing factor 3b associates with both the U2 and U11/U12 small nuclear ribonucleoprotein complexes (U2 snRNP) of spliceosomes. This 14 kDa protein interacts directly with subunit 1 of the splicing factor 3b complex. This 14 kDa protein also interacts directly with the adenosine that carries out the first transesterification step of splicing at the pre-mRNA branch site.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF3B14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SF3B6
Gene Accession No. P59708, Q9Y3B4
Gene Alias 0610009D07Rik; 6030419K15Rik; AV001342; CGI-110; HSPC175; Ht006; P14; pre mRNA branch site protein p14; pre-mRNA branch site protein p14; SAP14; SAP14a; SF3b 14 kDa subunit; Sf3b14; SF3B14a; SF3B6; spliceosome-associated protein, 14 kDa subunit; spliceosome-associated protein, 14-kDa; Splicing factor 3B subunit 6; splicing factor 3B, 14 kDa subunit; splicing factor 3B, 14 kDa subunit-like; splicing factor 3B, subunit 6; splicing factor 3b, subunit 6, 14kDa
Gene Symbols SF3B6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human SF3B6 (NP_0571311).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51639, 66055
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.