Learn More
SF3b145/SAP145 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00317

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Pig, Panda, Monkey, Gorilla, Chimpanzee, White-tufted-ear marmos
Subunit of the splicing factor SF3B required for ′A′ complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA. May also be involved in the assembly of the ′E′ complex. Belongs also to the minor U12-dependent spliceosome, which is involved in the splicing of rare class of nuclear pre-mRNA intron.Specifications
SF3b145/SAP145 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q13435 | |
Sf3b2 | |
Between 775 and 825 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
SF3B2 | |
Splicing factor 3B subunit 2, Pre-mRNA-splicing factor SF3b 145 kDa subunit, SF3b145, SF3b150, Spliceosome-associated protein 145, SAP 145, SF3B2, SAP145 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10992, 319322 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.