Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF3B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596456 Supplier Invitrogen™ Supplier No. PA596456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLGMPVGPNA HKVPPPWLIA MQRYGPPPSY PNLKIPGLNS PIPESCSFGY HAGGWGKPPV DETGKPLYGD VFGTNAAEFQ TKTEEEEIDR TPWGELEPSD EESSEEEEEE ESDEDKPDET GFITPADSGL ITPGGFSSVP AGMETPELIE LRKKKIEEAM DGSETPQLFT VLPEKRTATV GGAMMGSTHI YDMSTVMSRK GPAPELQGVE VALAPEELEL DPMAMTQKYE EHVREQQAQV EKEDFSDMVA EHAAKQKQKK RKAQPQDSRG GSKKYKEFKF; Positive Samples: HeLa, MCF7, SW620, A375, Mouse testis; Cellular Location: Nucleus.

Subunit of the splicing factor SF3B required for 'A' complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA. May also be involved in the assembly of the 'E' complex. Belongs also to the minor U12-dependent spliceosome, which is involved in the splicing of rare class of nuclear pre-mRNA intron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF3B2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SF3B2
Gene Accession No. Q13435
Gene Alias 145kDa; 2610311M13Rik; 2810441F20Rik; B230398H18Rik; Cus1; pre-mRNA splicing factor SF3b 145 kDa subunit; pre-mRNA-splicing factor SF3b 145 kDa subunit; SAP 145; SAP145; SF3b1; SF3B145; SF3b150; SF3B2; spliceosome associated protein 145; spliceosome-associated protein 145; Splicing factor 3B subunit 2; splicing factor 3b, subunit 2; splicing factor 3b, subunit 2, 145kD; splicing factor 3b, subunit 2, 145kDa
Gene Symbols SF3B2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 616-895 of human SF3B2 (NP_0068332).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10992, 293671, 319322
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.