Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF3B3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110358 Supplier Invitrogen™ Supplier No. PA5110358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPAAIAPFQG RVLIGVGKLL RVYDLGKKKL LRKCENKHIA NYISGIQTIG HRVIVSDVQE SFIWVRYKRN ENQLIIFADD TYPRWVTTAS LLDYDTVAGA DKFGNICVVR LPPNTNDEVD EDPTGNKALW DRGLLNGASQ KAEVIMNYHV GETVLSLQKT TLIPGGSESL VYTTLSGGIG ILVPFTSHED HDFFQHVEMH LRSEHPPLCG RDHLSFRSYY FPVKNVIDGD LCEQFNSMEP NKQKNVSEEL DRTPPEVSKK LEDIRTRYAF.

This gene encodes subunit 3 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex transcription coactivator-HAT complex, and the TFTC -containing complex). These complexes may function in chromatin modification, transcription, splicing, and DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF3B3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SF3B3
Gene Accession No. Q15393, Q921M3
Gene Alias 1810061H24Rik; 5730409A01Rik; AA409318; D8Ertd633e; Kiaa0017; mKIAA0017; pre-mRNA splicing factor SF3b, 130 kDa subunit; pre-mRNA-splicing factor SF3b 130 kDa subunit; RSE1; SAP 130; SAP130; SF3b130; SF3B3; Spliceosome-associated protein 130; splicing factor 3b subunit 3; splicing factor 3b, subunit 3; STAF130
Gene Symbols SF3B3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 948-1217 of human SF3B3/SAP130 (NP_0365583).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101943, 23450, 292019
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.