Learn More
SF3B3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | SF3B3 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2582
![]() |
Bethyl Laboratories, Inc
IHC00711T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2581
![]() |
Bethyl Laboratories, Inc
IHC00711 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_101
This gene encodes subunit 3 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex transcription coactivator-HAT complex, and the TFTC -containing complex). These complexes may function in chromatin modification, transcription, splicing, and DNA repair.Specifications
SF3B3 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
SF3B3; SAP130; splicing factor 3b, subunit 3, 130kDa; RSE1; SF3B130; KIAA0017; STAF130; pre-mRNA splicing factor SF3b, 130 kDa subunit; spliceosome-associated protein 130; splicing factor 3b, subunit 3; SF3b130 | |
Between 1167 and 1217 | |
Primary | |
4° C | |
SF3B3 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q15393, Q921M3 | |
101943, 23450 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.