Learn More
SF3B4 Antibody (3A1), DyLight 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP192692V
Catalog No. NBP192692V
Only null left
Add to Cart
Description
Description
SF3B4 Monoclonal specifically detects SF3B4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SF3B4
Monoclonal
1mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Hsh49, pre-mRNA splicing factor SF3b 49 kDa subunit, Pre-mRNA-splicing factor SF3b 49 kDa subunit, SAP 49, SAP49MGC10828, SF3B49, SF3b50, spliceosomal protein, spliceosome-associated protein (U2 snRNP), Spliceosome-associated protein 49, splicing factor 3B subunit 4, splicing factor 3b, subunit 4, 49kD, splicing factor 3b, subunit 4, 49kDa
Mouse
49 kDa
0.1 mL
Neuroscience
10262
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
3A1
DyLight 405
Q15427
SF3B4
Full length recombinant human SF3B4 expressed in and purified from E. coli. [UniProt# Q15427]
Protein G purified
RUO
Primary
Human, Bovine, Porcine, Mouse, Rat and other Mammals.
Store at 4C in the dark.
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only