Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF3B4 Monoclonal Antibody (3A1)
GREENER_CHOICE

Catalog No. PIMA547692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547692 Supplier Invitrogen™ Supplier No. MA547692
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specific for endogenous levels of the ~49 kDa SAP49 protein.

This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF3B4
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5 mM sodium azide
Gene Sf3b4
Gene Accession No. Q15427, Q6AYL5, Q8QZY9
Gene Alias 49kD; AFD1; chunp6902; fb36a04; Hsh49; MGC10828; pre-mRNA-splicing factor SF3b 49 kDa subunit; SAP 49; Sap49; SAP49MGC10828; sf3b4; SF3b49; SF3b50; spliceosomal protein; spliceosome-associated protein (U2 snRNP); spliceosome-associated protein 49; Splicing factor 3B subunit 4; splicing factor 3b, subunit 4; splicing factor 3b, subunit 4, 49kD; splicing factor 3b, subunit 4, 49kDa; Unknown (protein for IMAGE:8120437); wu:fb36a04
Gene Symbols Sf3b4
Host Species Mouse
Immunogen Recombinant full length human SAP49 expressed in and purified from E. Coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10262, 107701, 295270
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.