Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SF3B4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110359
Description
Immunogen sequence: MAAGPISERN QDATVYVGGL DEKVSEPLLW ELFLQAGPVV NTHMPKDRVT GQHQGYGFVE FLSEEDADYA IKIMNMIKLY GKPIRVNKAS AHNKNLDVGA NIFIGNLDPE IDEKLLYDTF SAFGVILQTP KIMRDPDTGN SKGYAFINFA SFDASDAAIE AMNGQYLCNR PITVSYAFKK DSKGERHGSA AERLLAAQNP.
This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.
Specifications
SF3B4 | |
Polyclonal | |
Unconjugated | |
Sf3b4 | |
49kD; AFD1; chunp6902; fb36a04; Hsh49; MGC10828; pre-mRNA-splicing factor SF3b 49 kDa subunit; SAP 49; Sap49; SAP49MGC10828; sf3b4; SF3b49; SF3b50; spliceosomal protein; spliceosome-associated protein (U2 snRNP); spliceosome-associated protein 49; Splicing factor 3B subunit 4; splicing factor 3b, subunit 4; splicing factor 3b, subunit 4, 49kD; splicing factor 3b, subunit 4, 49kDa; Unknown (protein for IMAGE:8120437); wu:fb36a04 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10262, 107701, 295270 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.86 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q15427, Q6AYL5, Q8QZY9 | |
Sf3b4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human SF3B4 (NP_005841.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction