Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112709 Supplier Thermo Scientific Supplier No. PA5112709
Add to Cart
Add to Cart

SFI1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen SFI1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Sfi1
Gene Accession No. A8K8P3
Gene Alias homolog of yeast Sfi1; hSFI1; hSfi1p; KIAA0542; PISD; PPP1R139; protein phosphatase 1, regulatory subunit 139; protein SFI1 homolog; Sfi1; SFI1 centrin binding protein; Sfi1 homolog, spindle assembly associated; Sfi1 homolog, spindle assembly associated (yeast)
Gene Symbols Sfi1
Host Species Rabbit
Immunogen Recombinant Human Protein SFI1 homolog protein (1003-1242AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9814
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.