Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ sfl Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144403 Supplier Invitrogen™ Supplier No. PA5144403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

sfl Polyclonal Antibody for Western Blot, ELISA

Essential bifunctional enzyme that catalyzes both the N-deacetylation and the N-sulfation of glucosamine (GlcNAc) of the glycosaminoglycan in heparan sulfate. Modifies the GlcNAc-GlcA disaccharide repeating sugar backbone to make N-sulfated heparosan, a prerequisite substrate for later modifications in heparin biosynthesis. Plays a role in diffusion of morphogen wingless (wg) via its role in heparan sulfate proteoglycans (HSPGs) biosynthesis, HSPGs being required for movement of wg morphogens. Required for wg signaling during both embryonic and imaginal disk development. Also required for FGF receptor signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen sfl
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.032 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene sfl
Gene Accession No. Q9V3L1
Gene Alias Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase
Gene Symbols sfl
Host Species Rabbit
Immunogen Recombinant Drosophila melanogaster (Fruit fly) NDST protein (1-172aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 38736
Target Species Drosophila
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.