Learn More
Description
- ADCF
- Supports high transfection efficiency
- Metabolically designed for high cell yield and recombinant protein/vector production
- Allows for direct or sequential adaptation
- Component traceability
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications
Specifications
| Form | Liquid |
| Product Type | SFM4Transfx-293 |
| Without Additives | No Glutamine, No Animal Derived Components |
| Quantity | 6 x 1000 mL |
| Certifications/Compliance | cGMP |
| Content And Storage | 2° to 8°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.