Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SFM4Transfx-293 Media

Catalog No. p-7058081
Click to view available options
Quantity:
1000 mL
6 x 1000 mL

Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.

  • ADCF
  • Supports high transfection efficiency
  • Metabolically designed for high cell yield and recombinant protein/vector production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type SFM4Transfx-293
Sterilization Method 0.2 μm Sterile-filtered
With Additives Poloxamer 188, Sodium Bicarbonate
Without Additives No Protein, No Phenol Red, No HEPES, No Glutamine
Quantity 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.