Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFMBT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584654 Supplier Invitrogen™ Supplier No. PA584654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YTHYYGKKKN KRIGRPPGGH SNLACALKKA SKRRKRRKNV FVHKKKRSSA SVDNTPAGSP QGS Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.

This gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. It encodes a protein which contains four malignant brain tumor repeat (mbt) domains and may be involved in antigen recognition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFMBT1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SFMBT1
Gene Accession No. Q9UHJ3
Gene Alias 4930442N21Rik; 9330180L21Rik; AA536974; hSFMBT; Renal ubiquitous protein 1; RU1; Scm like with four mbt domains 1; Scm-like with four mbt domains 1; scm-like with four MBT domains protein 1; Scm-related gene containing four mbt domains; Scm-related gene containing four mbt domains 1; Scm-related gene product containing four mbt domains; SFMBT; SFMBT1; Smr; wu:fc12f05
Gene Symbols SFMBT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SFMBT1. Recombinant protein control fragment (Product #RP-105319).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51460
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.