Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFMBT1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993677
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3677 100 μL
50-199-3678 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3677 Supplier Active Motif Supplier No. 61759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. It encodes a protein which contains four malignant brain tumor repeat (mbt) domains and may be involved in antigen recognition.

Specifications

Antigen SFMBT1
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene SFMBT1
Gene Accession No. Q9JMD1
Gene Alias 4930442N21Rik; 9330180L21Rik; AA536974; BOS_21049; hSFMBT; Renal ubiquitous protein 1; RU1; Scm like with four mbt domains 1; Scm-like with four mbt domains 1; Scm-like with four MBT domains protein 1; Scm-related gene containing four mbt domains; Scm-related gene containing four mbt domains 1; Scm-related gene product containing four mbt domains; Sfmbt; SFMBT1; Smr; wu:fc12f05
Gene Symbols SFMBT1
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the C-terminal region of mouse SFMBT1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54650
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.