Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFMBT1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564576
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4576 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4576 Supplier Bethyl Laboratories, Inc Supplier No. A303221AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat

SFMBT1 (Scm-like with four MBT domains 1) gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. It encodes a protein which contains four maligt brain tumor repeat (MBT) domains, and may be involved in antigen recognition. Several alternative splice variants that encode the same protein have been characterized.

Specifications

Antigen SFMBT1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SFMBT1
Gene Accession No. Q9UHJ3
Gene Alias RU1, SFMBT, Scm-related gene containing four mbt domains, Scm-related gene product containing four mbt domains, renal ubiquitous protein 1, scm-like with four MBT domains protein 1
Gene Symbols SFMBT1
Host Species Rabbit
Immunogen Between 816 and 866
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51460
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.