Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFMBT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583272
Description
Immunogen sequence: DQWLFYLDYR LRPVGWCQEN KYRMDPPSEI YPLKMASEWK CTLEKSLIDA AKFPLPMEVF KDHADLRSHF FTVGMKLETV NMCEPF.
The SFMBT2 (Scm-like with four MBT domains 2) gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. Sfmbt2 is a Polycomb group (PcG) gene that maps to the proximal region of Chromosome 2, and is a putative imprinted gene. Sfmbt2 is the first imprinted gene within this region to be identified. Studies indicate that six different translocations involving proximal chromosome 2 results in lethality when present as a maternal uniparental duplication.
Specifications
SFMBT2 | |
Polyclonal | |
Unconjugated | |
SFMBT2 | |
D2Wsu23e; D330030P06Rik; KIAA1617; Scm-like with 4 MBT domains protein 2; Scm-like with four mbt domains 2; scm-like with four MBT domains protein 2; Scm-related gene containing four mbt domain 2; Scm-related gene containing four mbt domains 2; SFMBT2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57713 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VUG0 | |
SFMBT2 | |
Recombinant protein corresponding to Human SFMBT2. Recombinant protein control fragment (Product #RP-97215). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction