Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFPQ Recombinant Rabbit Monoclonal Antibody (JU33-99)
GREENER_CHOICE

Catalog No. PIMA534648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534648 Supplier Invitrogen™ Supplier No. MA534648
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: K562, Daudi, mouse testis tissue, mouse lung tissue lysate, rat brain tissue, human tonsil tissue, human liver cancer tissue, human kidney tissue.

SFPQ, also named PSF, encodes a nuclear factor implicated in the splicing and regulation of gene expression. SFPQ probably forms a heteromer with NONO and participates in DNA pairing and DNA break repair program. Very recently SFPQ was identified as a downstream target of tau, complete nuclear depletion and cytoplasmic accumulation of SFPQ were shown in the neurons and astrocytes of brains with Alzheimer's disease (AD), more strikingly, reduced SFPQ levels may progress together with tau pathology, these observation strongly suggests the important role of SFPQ pathology in neurodegenerative diseases including AD. SFPQ protein may appears as 47 kDa, 52 kDa or 100 kDa molecules in various cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFPQ
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JU33-99
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SFPQ
Gene Accession No. P23246
Gene Alias 100 kDa DNA-pairing protein; 1110004P21Rik; 2810416M14Rik; 5730453G22Rik; 9030402K04Rik; AU021830; D4Ertd314e; DNA-binding p52/p100 complex, 100 kDa subunit; Gm12940; hPOMp100; NonO/p54nrb homolog; OTTMUSG00000009329; polypyrimidine tract binding protein associated; polypyrimidine tract-binding protein-associated splicing factor; polypyrimidine tract-binding protein-associated-splicing factor; POMP100; PPP1R140; protein phosphatase 1, regulatory subunit 140; PSF; PTB-associated splicing factor; PTB-associated-splicing factor; REP1; Sfpq; splicing factor proline and glutamine rich; splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated); splicing factor proline/glutamine rich (polypyrimidine tract-binding protein-associated); splicing factor proline/glutamine-rich; splicing factor, proline- and glutamine-rich
Gene Symbols SFPQ
Host Species Rabbit
Immunogen Synthetic peptide within Human SFPQ aa 658-707.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6421
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.