Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sFRP-1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299447IR
Description
sFRP-1 Polyclonal specifically detects sFRP-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
sFRP-1 | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1 | |
Produced in rabbits immunized with purified, recombinant sFRP-1 (NP_003003.3; Met1-Lys314). | |
0.1 mL | |
Apoptosis | |
6422 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction