Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sFRP-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | sFRP-1 |
---|---|
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
sFRP-1 Polyclonal antibody specifically detects sFRP-1 in Human samples. It is validated for ELISASpecifications
sFRP-1 | |
ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis | |
0.2 um filtered solution in PBS | |
6422 | |
IgG | |
Antigen and protein A Affinity-purified |
ELISA 1:5000-1:10000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1 | |
Produced in rabbits immunized with purified, recombinant Human sFRP-1 (Accession#: NP_003003.3; Met1-Lys314) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title