Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sFRP-1 Rabbit anti-Human, Mouse, Rat, Clone: 5I6V1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316851100UL
Description
sFRP-1 Monoclonal antibody specifically detects sFRP-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
sFRP-1 | |
Monoclonal | |
Unconjugated | |
FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human sFRP-1 (Q8N474). | |
100 μg | |
Apoptosis | |
6422 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
5I6V1 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction