Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sFRP-2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318244B
Description
sFRP-2 Polyclonal antibody specifically detects sFRP-2 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
sFRP-2 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2 | |
Synthetic peptide from the C-terminal of Human Secreted frizzled-related protein 2 (aa. 195-295) | |
0.1 mL | |
Stem Cell Markers | |
6423 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Biotin | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction