Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sFRP-2 Antibody (OTI6C2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274151R
Description
sFRP-2 Monoclonal specifically detects sFRP-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
sFRP-2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SFRP2 | |
Recombinant protein fragment corresponding to amino acids 25-295 of human sFRP-2 (NP_003004) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Immunohistochemistry | |
OTI6C2 | |
Immunohistochemistry | |
FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
6423.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction