Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRP2 Mouse anti-Human, Rat, Clone: 1F3A1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SFRP2 |
---|---|
Clone | 1F3A1 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6760
![]() ![]() |
Proteintech Group Inc
663281IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer.Specifications
SFRP2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FRP 2, FRP2, SARP 1, SARP1, SDF 5, sFRP 2, SFRP2, UNQ361/PRO697 | |
SFRP2 | |
IgG1 | |
Protein G | |
Antibody |
1F3A1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
Q96HF1 | |
310552, 6423 | |
SFRP2 Fusion Protein Ag18840 | |
Primary | |
-20°C | |
SFRP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title