Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRP2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA518656
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518656 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518656 Supplier Thermo Scientific Supplier No. PA518656

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology.

SFRP2 Polyclonal antibody specifically detects SFRP2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SFRP2
Gene Accession No. Q96HF1
Gene Alias AI851596; FKSG12; FRP 2; FRP2; FRP-2; MGC128911; MGC153618; MGC53344; Protein SDF5; SARP 1; SARP1; SARP-1; SDF 5; Sdf5; SDF-5; secreted apoptosis related protein 1; secreted apoptosis-related protein 1; secreted frizzled related protein 2; secreted frizzled-related protein 2; secreted frizzled-related sequence protein 2; secreted frizzled-related sequence protein 5; sFRP 2; SFRP2; sFRP-2; Sfrp2 protein; stromal cell derived factor 5; testicular tissue protein Li 170; UNQ361/PRO697
Gene Symbols SFRP2
Host Species Goat
Immunogen Synthetic peptide sequence (KRWQKGQREFKR) corresponding to the C-terminus amino acids of SFRP2
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6423
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.