Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRP4 Recombinant Rabbit Monoclonal Antibody (7Z6Q1)
SFRP4 Recombinant Rabbit Monoclonal Antibody (ARC0923)
Supplier: Thermo Scientific MA535460
Description
SFRP4 Recombinant Monoclonal Antibody for Western Blot

Specifications
SFRP4 | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6FHJ7, Q9JLS4, Q9Z1N6 | |
SFRP4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 201-300 of human SFRP4 (Q6FHJ7). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
7Z6Q1 | |
Unconjugated | |
SFRP4 | |
Ddc4; DDC-4 protein; Frizzled protein, human endometrium; frizzled related protein; frizzled-related protein; frizzled-related protein 4; FRP; FRP-4; FRPHE; MGC26498; secreted frizzled related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related sequence protein 4; Sfrp4; sFRP-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20379, 6424, 89803 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction