Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110360 Supplier Invitrogen™ Supplier No. PA5110360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRGAPCEAVR IPMCRHMPWN ITRMPNHLHH STQENAILAI EQYEELVDVN CSAVLRFFLC AMYAPICTLE FLHDPIKPCK SVCQRARDDC EPLMKMYNHS WPESLACDEL PVYDRGVCIS PEAIVTDLPE DVKWIDITPD MMVQERPLDV DCKRLSPDRC KCKKVKPTLA TYLSKNYSYV IHAKIKAVQR SGCNEVTTVV DVKEIFKSSS PIPRTQVPLI TNSSCQCPHI LPHQDVLIMC YEWRSRMMLL ENCLVEKWRD QLSKRSIQWE ERLQEQRRTV QDKKKTAGRT SRSNPPKPKG KPPAPKPASP KKNIKTRSAQ KRTNPKRV.

Secreted Frizzled-Related Proteins (sFRPs) are a family of glycosylated Wnt antagonists characterized by a conserved cysteine-rich domain that shares homology with the cysteine-rich, extracellular domain Frizzled proteins use for the binding of Wnt proteins and receptors. Lacking the transmembrane and intracellular domains of the Frizzled proteins, sFRPs function as soluble modulators of the Wnt signaling pathway through the direct binding of Wnt proteins to this cysteine-rich domain, and the resultant inhibition of Wnt receptor binding and signaling capabilities. sFRP-4 is widely distributed in a variety of embryonic and adult tissues where it can function as a circulating antiangiogenic factor, a potent proapoptotic factor, an inhibitor of insulin secretion, and a suppressor of both tumor growth and metastatic potential through disruption of the Wnt signaling pathway. Research has demonstrated the existence of a direct correlation between the downregulation and/or absence of circulating sFRP-4 and the progression of several cancer types, including ovarian, endometrial, prostate and lung. Upregulation of circulating sFRP-4 has been linked to the deterioration of glucose metabolism in the case of type 2 diabetes, as well as the suppression of the keratinocyte hyperproliferation and epidermal hyperlasia that are definitive of psoriasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRP4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SFRP4
Gene Accession No. Q6FHJ7, Q9JLS4, Q9Z1N6
Gene Alias Ddc4; DDC-4 protein; Frizzled protein, human endometrium; frizzled related protein; frizzled-related protein; frizzled-related protein 4; FRP; FRP-4; FRPHE; MGC26498; secreted frizzled related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related sequence protein 4; Sfrp4; sFRP-4
Gene Symbols SFRP4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 19-346 of human SFRP4 (NP_003005.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20379, 6424, 89803
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.