Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRS10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584604 Supplier Invitrogen™ Supplier No. PA584604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYDRGYDDRD YYSRSYRGGG GGG Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRA2B
Gene Accession No. P62995
Gene Alias 5730405G21Rik; D16Ertd266e; Htra2-beta; PPP1R156; protein phosphatase 1, regulatory subunit 156; RA301; serine/arginine-rich splicing factor 10; SFRS10; SIG-41; Silg41; Silica-induced gene 41 protein; similar to splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila); splicing factor, arginine/serine-rich 10; splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila); splicing factor, arginine/serine-rich, 10; SRFS10; Srsf10; TRA-2 beta; Tra2b; TRA2beta; TRA2-beta; TRAN2B; transformer 2 beta homolog; transformer 2 beta homolog (Drosophila); transformer-2 protein homolog B; Transformer-2 protein homolog beta; zgc:56612
Gene Symbols TRA2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRA2B. Recombinant protein control fragment (Product #RP-103845).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6434
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.