Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRS12 Monoclonal Antibody (1D11)
GREENER_CHOICE

Catalog No. PIMA523061
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523061 Supplier Invitrogen™ Supplier No. MA523061
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: NDRDGKEKLE KNSTSLKEKE HNKEPDSSVS KEVDDKDAPR TEENKIQHNG NCQLNEENLS TKTEAV.

SFRS12 belongs to the superfamily of serine/arginine-rich splicing factors. It modulates splice site selection by regulating the activities of other SR proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS12
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SREK1
Gene Accession No. Q8WXA9
Gene Alias 8430401B01; AI450757; AI462342; serine/arginine-rich splicing factor 12; serine/arginine-rich-splicing regulatory protein 86; serine-arginine-rich splicing regulatory protein 508; serine-arginine-rich splicing regulatory protein 86; serine-arginine-rich-splicing regulatory protein 86; SFRS12; Splicing factor, arginine/serine-rich 12; splicing regulatory glutamic acid and lysine rich protein 1; splicing regulatory glutamic acid/lysine-rich protein 1; splicing regulatory glutamine/lysine-rich protein 1; Splicing regulatory protein 508; Srek1; SR-related protein of 86 kDa; SRrp508; Srrp86; Srsf12
Gene Symbols SREK1
Host Species Mouse
Immunogen SREK1 (NP_631907, 443 a.a. ~ 508 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140890
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.