Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRS12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584412 Supplier Invitrogen™ Supplier No. PA584412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KERDHISERR ERERSTSMRK SSNDRDGKEK LEKNSTSLKE KEHNKEPDSS VSKEVDDKDA PRTEENKIQH NGNCQLNEEN LSTKTEAV Highest antigen sequence indentity to the following orthologs: Rat - 69%, Mouse - 69%.

SFRS12 belongs to the superfamily of serine/arginine-rich splicing factors. It modulates splice site selection by regulating the activities of other SR proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS12
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SREK1
Gene Accession No. Q8WXA9
Gene Alias 8430401B01; AI450757; AI462342; serine/arginine-rich splicing factor 12; serine/arginine-rich-splicing regulatory protein 86; serine-arginine-rich splicing regulatory protein 508; serine-arginine-rich splicing regulatory protein 86; serine-arginine-rich-splicing regulatory protein 86; SFRS12; Splicing factor, arginine/serine-rich 12; splicing regulatory glutamic acid and lysine rich protein 1; splicing regulatory glutamic acid/lysine-rich protein 1; splicing regulatory glutamine/lysine-rich protein 1; Splicing regulatory protein 508; Srek1; SR-related protein of 86 kDa; SRrp508; Srrp86; Srsf12
Gene Symbols SREK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SREK1. Recombinant protein control fragment (Product #RP-106929).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.