Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRS15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553838 Supplier Invitrogen™ Supplier No. PA553838
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PITPPVSIPP PHTPPISIPN STIAGINEDT TKDLSIGNPI PTVVSGARGN AESGDSVKMY GSAVPPAAPT NLPTPPVTQP VSLLGTQGVA PGPVIGLQAP STGLLGARPG LIPLQRPPGM P Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 78%.

The family of SR factors all contain one or more RNA recognition motifs (RRM) and an SR-rich domain. They are not only essential for constitutive splicing, but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. Splicing factor arginine/serine-rich 15 (SFRS15), also designated CTD-binding SR-like protein RA4, contains one RRM and one SR-rich domains. SFRS15 interacts with C-terminal repetitive domain (CTD) of Pol II and is believed to functionally and physically link transcription and pre-mRNA processing. Localized to the nucleus, SFRS15 is expressed as two isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS15
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCAF4
Gene Accession No. O95104
Gene Alias AA517739; CTD-binding SR-like protein RA4; KIAA1172; pre-mRNA splicing SR protein rA4; SCAF4; serine/arginine-rich splicing factor 15; SFR15; SFRS15; splicing factor serine alanine 15; splicing factor, arginine/serine-rich 15; SRA4; SR-like CTD-associated factor 4; SR-related and CTD-associated factor 4; SR-related CTD associated factor 4; SR-related CTD-associated factor 4; Srsf15
Gene Symbols SCAF4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SFRS15. Recombinant protein control fragment (Product #RP-91821).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57466
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.