Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS17A Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB29955
Description
Rabbit polyclonal antibody raised against synthetic peptide of human SFRS17A.
- High specificity
- High reactivity
Specifications
SFRS17A | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of human SFRS17A. | |
In PBS (2% sucrose, 0.09% sodium azide). | |
721P/CCDC133/CXYorf3/DXYS155E/MGC125365/MGC125366/MGC39904/XE7/XE7Y | |
Rabbit | |
Affinity purification | |
RUO | |
8227 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
NM_005088 | |
SFRS17A | |
A synthetic peptide corresponding to N-terminus of human SFRS17A. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction