Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRS18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568338 Supplier Invitrogen™ Supplier No. PA568338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human and Rat samples.

SFRS18 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS18
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PNISR
Gene Accession No. A2AJT4, Q8TF01
Gene Alias 5730406M06Rik; AA673174; argi; arginine/serine-rich protein PNISR; bA98I9.2; C6orf111; HSPC261; HSPC306; Pinsr; PNISR; PNN interacting serine and arginine rich protein; PNN interacting serine/arginine-rich; PNN-interacting serine/arginine-rich protein; RGD1307395; RP11-98I9.2; serine/arginine-rich splicing factor 18; serine/arginine-rich-splicing regulatory protein 130; serine-arginine-rich-splicing regulatory protein 130; Sfrs18; splicing factor, arginine/serine-rich 130; Splicing factor, arginine/serine-rich 18; SR rich protein; SR-related protein; SR-rich protein; SRrp130; Srsf18
Gene Symbols PNISR
Host Species Rabbit
Immunogen Synthesized peptide derived from human SFRS18.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25957, 297942, 66625
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.